Merck
  • Cutinase-like enzyme from the yeast Cryptococcus sp. strain S-2 hydrolyzes polylactic acid and other biodegradable plastics.
Cutinase-like enzyme from the yeast Cryptococcus sp. strain S-2 hydrolyzes polylactic acid and other biodegradable plastics.
Applied and environmental microbiology (2005-11-05)
Kazuo Masaki, Numbi Ramudu Kamini, Hiroko Ikeda, Haruyuki Iefuji
ABSTRACT
A purified lipase from the yeast Cryptococcus sp. strain S-2 exhibited remote homology to proteins belonging to the cutinase family rather than to lipases. This enzyme could effectively degrade the high-molecular-weight compound polylactic acid, as well as other biodegradable plastics, including polybutylene succinate, poly (epsilon-caprolactone), and poly(3-hydroxybutyrate).
Sigma-Aldrich
Polybutenes, average Mn ~2,300 by VPO, isobutylene >90 %
Sigma-Aldrich
Polybutenes, average Mn ~920 by VPO, isobutylene >90 %
Sigma-Aldrich
Proteinase K, (from Tritirachium album) chromatographically purified lyophilized 30 mAnson-U/mg for biochemistry and molecular biology EC 3.4.21.14