Merck
  • Selective synthesis of C-arylglycosides via Cp*RuCl-catalyzed partially intramolecular cyclotrimerizations of C-alkynylglycosides.
Selective synthesis of C-arylglycosides via Cp*RuCl-catalyzed partially intramolecular cyclotrimerizations of C-alkynylglycosides.
Organic & biomolecular chemistry (2005-04-29)
Yoshihiko Yamamoto, Tomoaki Saigoku, Hisao Nishiyama, Takashige Ohgai, Kenji Itoh
ABSTRACT
In the presence of catalytic amounts of Cp*RuCl(cod), the partially intramolecular cyclotrimerizations of various C-alkynylglycosides and C-diynylglycosides proceeded at ambient temperature to afford C-arylglycosides.
Sigma-Aldrich
Ruthenium(III) chloride, Ru content 45-55%