Merck
  • Characterization of Anopheles gambiae transglutaminase 3 (AgTG3) and its native substrate Plugin.
Characterization of Anopheles gambiae transglutaminase 3 (AgTG3) and its native substrate Plugin.
The Journal of biological chemistry (2013-01-05)
Binh V Le, Jennifer B Nguyen, Shankar Logarajah, Bo Wang, Jacob Marcus, Hazel P Williams, Flaminia Catteruccia, Richard H G Baxter
ABSTRACT
Male Anopheles mosquitoes coagulate their seminal fluids via cross-linking of a substrate, called Plugin, by the seminal transglutaminase AgTG3. Formation of the "mating plug" by cross-linking Plugin is necessary for efficient sperm storage by females. AgTG3 has a similar degree of sequence identity (~30%) to both human Factor XIII (FXIII) and tissue transglutaminase 2 (hTG2). Here we report the solution structure and in vitro activity for the cross-linking reaction of AgTG3 and Plugin. AgTG3 is a dimer in solution and exhibits Ca(2+)-dependent nonproteolytic activation analogous to cytoplasmic FXIII. The C-terminal domain of Plugin is predominantly α-helical with extended tertiary structure and oligomerizes in solution. The specific activity of AgTG3 was measured as 4.25 × 10(-2) units mg(-1). AgTG3 is less active than hTG2 assayed using the general substrate TVQQEL but has 8-10× higher relative activity when Plugin is the substrate. Mass spectrometric analysis of cross-linked Plugin detects specific peptides including a predicted consensus motif for cross-linking by AgTG3. These results support the development of AgTG3 inhibitors as specific and effective chemosterilants for A. gambiae.
Sigma-Aldrich
Transglutaminase Assay Kit, sufficient for assays in two 96-well plates
Sigma-Aldrich
Transglutaminase from guinea pig liver, lyophilized powder, ≥1.5 units/mg protein
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg