Merck
  • Fluorescent detection of dipicolinic acid as a biomarker in bacterial spores employing terbium ion-coordinated magnetite nanoparticles.
Fluorescent detection of dipicolinic acid as a biomarker in bacterial spores employing terbium ion-coordinated magnetite nanoparticles.
Journal of hazardous materials (2021-01-03)
Thomas Myeongseok Koo, Min Jun Ko, Bum Chul Park, Myeong Soo Kim, Young Keun Kim
ABSTRACT
Anthrax is a bioterror agent because of its toxicity and the tolerance of its bacterial spores. Thus, researchers have attempted to develop various nanomaterials to detect dipicolinic acid (DPA), a biomarker of bacterial spores. Nanomaterials containing lanthanide ions have received considerable attention, owing to their potential to exhibit high sensitivity and selectivity in the detection of DPA via chelation with molecules. However, the fluorescent signals of the lanthanide complex are quenchable because the nanomaterials simultaneously absorb the excitation and emission light. For the precise detection of DPA, pure signals have to be obtained from the complex by alleviating the quenching effect of the nanomaterials. In this study, we develop a structure with terbium ion (Tb3+)-coordinated magnetite (Fe3O4) nanoparticle to detect DPA. Tb3+ can be detached from the magnetite during chelation with the DPA, and the complex can emit the unencumbered signals with improved detection limit through the application of a magnetic field. The detection system exhibits a significantly lower detection limit (5.4 nM) than the infectious dosage of anthrax (60 μM) with high selectivity and chemical stability. This study informs the improvement of detection limits via the separation of nanomaterials and lanthanide complex.
Sigma-Aldrich
2-(Diisopropylamino)ethyl methacrylate, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Sodium acrylate, 97%
Sigma-Aldrich
Terbium(III) acetate hydrate, 99.9% trace metals basis