Merck
  • Secreted stromal protein ISLR promotes intestinal regeneration by suppressing epithelial Hippo signaling.
Secreted stromal protein ISLR promotes intestinal regeneration by suppressing epithelial Hippo signaling.
The EMBO journal (2020-03-05)
Jiuzhi Xu, Yang Tang, Xiaole Sheng, Yuhua Tian, Min Deng, Sujuan Du, Cong Lv, Guili Li, Yuwei Pan, Yongli Song, Pengbo Lou, Yongting Luo, Yuan Li, Bing Zhang, Yanmei Chen, Zhanju Liu, Yingzi Cong, Maksim V Plikus, Qingyong Meng, Zhaocai Zhou, Zhengquan Yu
ABSTRACT
The Hippo-YAP signaling pathway plays an essential role in epithelial cells during intestinal regeneration and tumorigenesis. However, the molecular mechanism linking stromal signals to YAP-mediated intestinal regeneration and tumorigenesis is poorly defined. Here, we report a stroma-epithelium ISLR-YAP signaling axis essential for stromal cells to modulate epithelial cell growth during intestinal regeneration and tumorigenesis. Specifically, upon inflammation and in cancer, an oncogenic transcription factor ETS1 in stromal cells induces expression of a secreted protein ISLR that can inhibit Hippo signaling and activate YAP in epithelial cells. Deletion of Islr in stromal cells in mice markedly impaired intestinal regeneration and suppressed tumorigenesis in the colon. Moreover, the expression of stromal cell-specific ISLR and ETS1 significantly increased in inflamed mucosa of human IBD patients and in human colorectal adenocarcinoma, accounting for the epithelial YAP hyperactivation. Collectively, our findings provide new insights into the signaling crosstalk between stroma and epithelium during tissue regeneration and tumorigenesis.
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines