Merck
  • RP11-81H3.2 promotes gastric cancer progression through miR-339-HNRNPA1 interaction network.
RP11-81H3.2 promotes gastric cancer progression through miR-339-HNRNPA1 interaction network.
Cancer medicine (2020-02-14)
Fen-Rong Chen, Su-Mei Sha, Shen-Hao Wang, Hai-Tao Shi, Lei Dong, Dong Liu, Yan Cheng, Miao An, Yan Wang, Jun Zhang
ABSTRACT
Recent studies have demonstrated that various long non-coding RNAs (lncRNAs) participate in the gastric cancer (GC) development and metastasis. Some lncRNAs exert their regulatory function by interacting with microRNAs. Here we identified a novel lncRNA RP11-81H3.2 that was highly expressed in the GC tissue and cell lines. RP11-81H3.2 knockdown significantly inhibited the proliferation, migration, and invasion of GC cells. Mechanistically, we demonstrated that RP11-81H3.2 directly interacted with miR-339 while miR-339 regulated the HNRNPA1 expression by targeting HRRNPA1 3'-UTR. RP11-81H3.2-miR-339-HNRNPA1 interaction network regulated the GC cell proliferation, migration, and invasion. Moreover, our results confirmed that RP11-81H3.2 knockdown suppressed the tumor growth of GC in a xenograft model in vivo. In summary, the results suggest that RP11-81H3.2 functions as an oncogene in GC and could be utilized as a promising diagnosis and therapeutic marker for GC treatment.