Merck
  • Recombinant Erythropoietin Provides Protection against Renal Fibrosis in Adenine-Induced Chronic Kidney Disease.
Recombinant Erythropoietin Provides Protection against Renal Fibrosis in Adenine-Induced Chronic Kidney Disease.
Mediators of inflammation (2020-03-19)
Estefanía Vázquez-Méndez, Yanet Gutiérrez-Mercado, Edgar Mendieta-Condado, Francisco Javier Gálvez-Gastélum, Hugo Esquivel-Solís, Yadira Sánchez-Toscano, Claudia Morales-Martínez, Alejandro A Canales-Aguirre, Ana Laura Márquez-Aguirre
ABSTRACT
Chronic kidney disease (CKD) causes anemia by renal damage. In CKD, the kidney is submitted to hypoxia, persistent inflammation, leading to fibrosis and permanent loss of renal function. Human recombinant erythropoietin (rEPO) has been widely used to treat CKD-associated anemia and is known to possess organ-protective properties that are independent from its well-established hematopoietic effects. Nonhematopoietic effects of EPO are mediated by an alternative receptor that is proposed to consist of a heterocomplex between the erythropoietin receptor (EPOR) and the beta common receptor (βcR). The present study explored the effects of rEPO to prevent renal fibrosis in adenine-induced chronic kidney disease (Ad-CKD) and their association with the expression of the heterodimer EPOR/βcR. Male Wistar rats were randomized to control group (CTL), adenine-fed rats (Ad-CKD), and Ad-CKD with treatment of rEPO (1050 IU/kg, once weekly for 4 weeks). Ad-CKD rats exhibited anemia, uremia, decreased renal function, increased infiltration of inflammatory cells, tubular atrophy, and fibrosis. rEPO treatment not only corrected anemia but reduced uremia and partially improved renal function as well. In addition, we observed that rEPO diminishes tubular injury, prevents fibrosis deposition, and induces the EPOR/βcR heteroreceptor. The findings may explain the extrahematopoietic effects of rEPO in CKD and provide new strategies for the treatment of renal fibrosis in CKD.
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
RIPA Buffer