Merck
  • Determination of 5-nitro-2-furaldehyde as marker residue for nitrofurazone treatment in farmed shrimps and with addressing the use of a novel internal standard.
Determination of 5-nitro-2-furaldehyde as marker residue for nitrofurazone treatment in farmed shrimps and with addressing the use of a novel internal standard.
Scientific reports (2019-12-19)
Qiang Wang, Xu-Feng Wang, Yong-Yuan Jiang, Zhi-Guang Li, Nan Cai, Wan-Qi Guan, Ke Huang, Dong-Hao Zhao
ABSTRACT
We developed a significantly improved ultra-high performance liquid chromatography-tandem mass spectrometry method for determination of 5-nitro-2-furaldehyde (NF) as a surrogate using a novel internal standard for the detection of nitrofurazone. We used 2,4-dinitrophenylhydrazine derivatization and furfural as the internal standard. Derivatization was easily performed in HCl using ultrasonic manipulation for 5 min followed by liquid extraction using ethyl acetate. The samples were concentrated and purified using reverse phase and alumina cartridges in tandem. The derivatives were separated using a linear gradient elution on a C18 column with methanol and water as the mobile phase in negative ionization mode and multiple reaction monitoring. Under the optimized conditions, the calibration curves were linear from 0.2 to 20 μg/L with correlation coefficients >0.999. Mean recoveries were 80.8 to 104.4% with the intra- and inter-day relative standard deviations <15% at spiking levels of 0.1 to 10 μg/kg. The limits of detection and quantification were 0.05 and 0.1 μg/kg, respectively. This method is a robust tool for the identification and quantitative determination of NF in shrimp samples.
Sigma-Aldrich
Semicarbazide, 6 wt. % (on silica gel)