Merck
  • Vasodilating effect of di-peptides in thoracic aortas from spontaneously hypertensive rats.
Vasodilating effect of di-peptides in thoracic aortas from spontaneously hypertensive rats.
Bioscience, biotechnology, and biochemistry (2006-09-09)
Mitsuru Tanaka, Toshiro Matsui, Yusuke Ushida, Kiyoshi Matsumoto
ABSTRACT
In this study, we found that antihypertensive di-peptide Val-Tyr (VY) showed a vascular relaxation effect in KCl-induced contraction of thoracic aorta rings from 18-week-old spontaneously hypertensive rats among di-peptides of VY, Ile-Tyr, and Tyr-Val irrespective of their angiotensin I-converting enzyme inhibitory activity. The effect was endothelium-independent, and was closely associated with vascular responses in the vascular smooth muscle layer.