Skip to Content
Merck
Ciprofloxacin hydrochloride for peak identification European Pharmacopoeia (EP) Reference Standard

Ciprofloxacin HCl

Ciprofloxacin hydrochloride for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Ciprofloxacin hydrochloride monohydrate
Empirical Formula (Hill Notation):
C17H18FN3O3 · HCl · H2O
CAS Number:
Molecular Weight:
385.82

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique