Camptothecin
Synonym(s):
(S)-(+)-Camptothecin
Empirical Formula (Hill Notation):
C20H16N2O4
CAS Number:
Molecular Weight:
348.35
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3