Sodium phosphate dibasic solution
Synonym(s):
Disodium hydrogen phosphate solution, di-Sodium hydrogen phosphate solution, Sodium phosphate dibasic solution
Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2