Polyethylene Glycol 1500
Synonym(s):
Poly(ethylene glycol), PEG, PEG solution, Polyethylene glycol solution
Linear Formula:
H(OCH2CH2)nOH
CAS Number:
Products
Product Category
Brand
Manufacturer
pH Value
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing