Skip to Content
Merck
5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine AldrichCPR

5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine

5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine AldrichCPR
Synonym(s):
7-Azaindole-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
Empirical Formula (Hill Notation):
C13H17BN2O2
CAS Number:
Molecular Weight:
244.10

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1