Skip to Content
Merck
p-Toluenesulfonic acid monohydrate ≥99% (calc. to H2O free subst.)

p-Toluenesulfonic acid monohydrate

p-Toluenesulfonic acid monohydrate ≥99% (calc. to H2O free subst.)
Synonym(s):
Toluene-4-sulfonic acid monohydrate
Linear Formula:
CH3C6H5SO3H · H2O
CAS Number:
Molecular Weight:
172.20 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Markush Class
Markush Group