
(16a)-16-(Acetyloxy)-17-hydroxy-3,20-dioxopregna-1,4,9(11)-trien-21-yl acetate

Synonym(s):
(16a)-16-(Acetyloxy)-17-hydroxy-3,20-dioxopregna-1,4,9(11)-trien-21-yl acetate
Empirical Formula (Hill Notation):
C25H30O7
CAS Number:
Molecular Weight:
442.50
Products
Formula Weight
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing