Ammonium heptamolybdate tetrahydrate
Synonym(s):
Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate, Ammonium molybdate tetrahydrate
Linear Formula:
(NH4)6Mo7O24 · 4H2O
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Analyte Suitability
Quality Segment
Application
Reagent Type
Core
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing