Skip to Content
Merck
Sodium carbonate-12C 99.9 atom % 12C

Sodium carbonate

Sodium carbonate-12C 99.9 atom % 12C
Synonym(s):
Calcined soda, Carbonic acid disodium salt, Soda ash, Sodium carbonate
Linear Formula:
Na212CO3
CAS Number:
Molecular Weight:
105.98
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Special Grade
Showing 1-20 of 20
Compare
Product Number
Description
Pricing