Merck
GC Analysis of QA Test Mix on SLB®-5ms, Three Different Phase Lots
GC Analysis of QA Test Mix on SLB®-5ms, Three Different Phase Lots suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
110 °C (14 min), 15 °C/min to 325 °C (15 min)
inj. temp.
250 °C
detector
FID, 360 °C
carrier gas
helium, 30 cm/sec @ 110 °C
injection
1.0 μL 100:1 split
liner
4 mm I.D. split, cup design
sample
Nonpolar Column Test Mix, each analyte at 500 μg/mL in methylene chloride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany