Merck
  • Identification and characterization of an alternative cytotoxic T lymphocyte-associated protein 4 binding molecule on B cells.
Identification and characterization of an alternative cytotoxic T lymphocyte-associated protein 4 binding molecule on B cells.
Proceedings of the National Academy of Sciences of the United States of America (1996-07-23)
M Murakami, Y Takahashi, Y Isashi, S Kon, W Y Jia, M Inobe, R Abe, T Uede
ABSTRACT
To determine whether alternative cytotoxic T lymphocyte-associated protein 4 (CTLA4) binding proteins exist on B cells, we constructed (i) mCTLA4hIgG consisting of the extracellular region of a mouse CTLA4 molecule and the Fc portion of a human IgG1 molecule and (ii) PYAAhIgG, a mutant mCTLA4hIgG, having two amino acid substitutions on the conserved MYPPPY motif in the complementarity-determining region 3-like region and lacking detectable binding to both B7-1 and B7-2 molecules. Using these fusion proteins (mCTLA4hIgG and PYAAhIgG), we demonstrated that a mouse immature B-cell line, WEHI231 cells, expressed alternative CTLA4 binding molecules (ACBMs) that were distinct from both B7-1 and B7-2. ACBMs were 130-kDa disulfide-linked proteins. More importantly, ACBMs were able to provide costimulatory signal for T-cell proliferation in the presence of anti-CD3 monoclonal antibodies. In addition, we demonstrated that more than 20% of B220+ cells obtained from normal mouse spleen expressed ACBMs.