Merck
  • High-performance liquid chromatographic determination of dyclonine hydrochloride in the presence of its degradation products.
High-performance liquid chromatographic determination of dyclonine hydrochloride in the presence of its degradation products.
Journal of pharmaceutical and biomedical analysis (1989-01-01)
H R Bhagat, H N Bhargava, D A Williams
ABSTRACT
A stability-indicating reversed-phase high-performance liquid chromatographic method for the determination of the topical anaesthetic dicyclonine hydrochloride in various oral spray and gel formulations is described.