Merck
  • The ecological effects of a herbicide-insecticide mixture on an experimental freshwater ecosystem.
The ecological effects of a herbicide-insecticide mixture on an experimental freshwater ecosystem.
Environmental pollution (Barking, Essex : 1987) (2012-10-20)
Catherine B Choung, Ross V Hyne, Mark M Stevens, Grant C Hose
ABSTRACT
The effects of a co-occurring insecticide-herbicide mixture were evaluated using model ecosystems (microcosms) in the laboratory. Microcosms dosed with a high concentration (10 μg/L) of the insecticide terbufos, alone and as a mixture with the herbicide atrazine (25 μg/L), exhibited community level effects attributed to the elimination and decline of invertebrate populations, and also indirect effects. There were no community level effects at a lower concentration of terbufos (0.1 μg/L) alone or in a mixture with atrazine, although delayed population effects were observed. Female chironomids also emerged later and those from terbufos-only microcosms were smaller. Exposure to atrazine alone was also associated with lower abundances of cladocerans and reduced emergence of chironomids. The risk posed by atrazine is low and is unlikely to exacerbate the effects of terbufos. Nevertheless, the population-level effects highlight that terbufos poses a potential risk to aquatic ecosystems, regardless of whether atrazine is also present.