Merck
  • Multiple fold increase in activity of ferroxidase-apoferritin complex by silver and gold nanoparticles.
Multiple fold increase in activity of ferroxidase-apoferritin complex by silver and gold nanoparticles.
Nanomedicine : nanotechnology, biology, and medicine (2012-07-10)
Afolake Sennuga, Jacqueline van Marwijk, Chris G Whiteley
ABSTRACT
The effect of silver (Ag) and gold (Au) nanoparticles on the ferroxidase activity of apoferritin showed a 110-fold increase in specific activity and a 9-fold increase over the control at the respective molar ratios of Au-apoferritin and Ag-apoferritin nanoparticles (NPs) of 500:1 and 1000:1. Typical color change, from pale yellow to brown, occurred when apoferritin was mixed with AgNO(3) or AuCl(3) followed by sodium borohydride to afford respective metal-apoferritin NP complexes in a ratio of between 250:1 and 4000:1. These complexes were characterized by ultraviolet-visible inductively coupled plasma-optical emission spectroscopy, Fourier transform infrared spectroscopy, transmission electron microscopy, and energy-dispersive x-ray spectroscopy. Transmission electron microscopy revealed that the size of NPs increased as the molar ratio of metal to apoferritin increased, with an average size of 3-6 nm generated with Au-to-apoferritin and/or Ag-to-apoferritin molar ratios of 250:1 to 4000:1. Fourier transform infrared spectrometry showed no structural changes of apoferritin when the NPs were attached to the protein. In this paper the utility of gold and silver nanoparticles in augmenting the activity of the ferroxidase-apoferritin complex is described. Both NPs dramatically increased the ferroxidase activity.