Merck
  • General approach to epipolythiodiketopiperazine alkaloids: total synthesis of (+)-chaetocins A and C and (+)-12,12'-dideoxychetracin A.
General approach to epipolythiodiketopiperazine alkaloids: total synthesis of (+)-chaetocins A and C and (+)-12,12'-dideoxychetracin A.
Journal of the American Chemical Society (2010-09-28)
Justin Kim, Mohammad Movassaghi
ABSTRACT
A highly stereoselective and systematic strategy for the introduction of polysulfides in the synthesis of epipolythiodiketopiperazines is described. We report the first total synthesis of dimeric epitri- and epitetrathiodiketopiperazines.
Sigma-Aldrich
Chaetocin from Chaetomium minutum, ≥95% (HPLC)