Merck
  • Effects of direct cedrol inhalation into the lower airway on autonomic nervous activity in totally laryngectomized subjects.
Effects of direct cedrol inhalation into the lower airway on autonomic nervous activity in totally laryngectomized subjects.
British journal of clinical pharmacology (2007-10-24)
Katsumi Umeno, Etsuro Hori, Masahito Tsubota, Hideo Shojaku, Takaki Miwa, Yoshinao Nagashima, Yukihiro Yada, Toshiyuki Suzuki, Taketoshi Ono, Hisao Nishijo
ABSTRACT
Relationships between smell sensation and autonomic changes have been studied extensively. However, the possibility that odorants may also act on the lung and lower airway remains unknown. The present results provide the first evidence that the lung and lower airway exert an inhibitory influence on the cardiovascular system in response to Cedrol (odorant) in the air under physiological conditions. Previous studies reported that Cedrol (odorant) inhalation (CI) induced changes in autonomic balance and baroreceptor sensitivity (BRS) in both healthy subjects and anosmic patients. This suggests that Cedrol may act on the lower airway, and that the pulmonary system may exert an inhibitory influence on the cardiovascular system. To test the above possibility, vaporized Cedrol (64.0 +/- 7.7 10(-9)m) or blank air was directly inhaled through the lower airway from a hole in the trachea, but not through the upper airway, using totally laryngectomized subjects. During the experiment, ECG, systolic (SBP) and diastolic (DBP) blood pressures were measured. Sympathetic and parasympathetic nervous activity was estimated by spectral analyses of variability in these parameters (heart rate variability (HRV), SBP variability (SBPV) and DBP variability (DBPV)). BRS was computed from transfer gain between SBP and the R-R interval of the ECG. SBP and DBP significantly decreased during CI, although there were no significant differences in HR and respiratory rate. BRS significantly increased during CI. The low frequency components of SBPV and DBPV (indices for sympathetic activity) significantly decreased during CI, while high frequency components of HRV (an index for parasympathetic activity) significantly increased. The present experiment using totally laryngectomized patients replicated the similar results in healthy subjects who inhaled Cedrol through the nose, suppression of sympathetic outflow and increase in parasympathetic outflow. These results demonstrated that Cedrol acts on the lower airway and pulmonary system, and suggest a new target for drug therapy of hypertension.
Sigma-Aldrich
(+)-Cedrol, ≥99.0% (sum of enantiomers, GC)