Merck
  • Comparison of ceftibuten transport across Caco-2 cells and rat jejunum mounted on modified Ussing chambers.
Comparison of ceftibuten transport across Caco-2 cells and rat jejunum mounted on modified Ussing chambers.
Biopharmaceutics & drug disposition (2003-10-02)
R M Menon, W H Barr
ABSTRACT
Ceftibuten uptake into Caco-2 cells and intestinal brush border membrane vesicles is mediated by the dipeptide transport system (PEPT1). The apical to basolateral transport characteristics of ceftibuten across Caco-2 cells and rat jejunum mounted on a modified Ussing chamber was examined. Mannitol was used as a paracellular marker along with trans-epithelial electrical resistance (TEER) for monitoring tight junction permeability. Transport across Caco-2 cells and rat jejunum mounted on a modified Ussing chamber was linear across the concentration range 0.25-10 mM. The net flux of mannitol and ceftibuten was higher across rat jejunum compared with Caco-2 cells. At a donor concentration of 0.25 mM, ceftibuten transport across Caco-2 cells was found to be pH dependent. Glycyl proline, a dipeptide, and 2,4- dinitrophenol, an energy poison, caused a reduction in the permeability of 0.25 mM ceftibuten across Caco-2 cells. Benzoic acid and adipic acid also inhibited transcellular transport of ceftibuten. At a donor concentration of 0.25 mM, passive paracellular transport accounts for about 60% and the active carrier mediated mechanism accounts for about 40% of ceftibuten transport across Caco-2 cells. None of the inhibitors however, had a significant effect on ceftibuten transport across rat jejunum mounted on a modified Ussing chamber at a donor concentration of 0.25 mM. In the concentration range 0.25-10 mM, ceftibuten is predominantly transported by paracellular mechanisms across rat jejunum and a mixture of active and passive transport across Caco-2 cells.
Sigma-Aldrich
Ceftibuten hydrate, ≥98% (HPLC)