- Characterization of the in vitro kinetic interaction of chlorpyrifos-oxon with rat salivary cholinesterase: a potential biomonitoring matrix.
Characterization of the in vitro kinetic interaction of chlorpyrifos-oxon with rat salivary cholinesterase: a potential biomonitoring matrix.
The primary mechanism of action for organophosphorus (OP) insecticides such as chlorpyrifos (CPF) involves the inhibition of acetylcholinesterase (AChE) by their active oxon metabolites resulting in a wide range of neurotoxic effects. These oxons also inhibit other cholinesterases (ChE) such as butyrylcholinesterase (BuChE), which represents a detoxification mechanism and a potential biomarker for OP insecticide exposure/response. Salivary biomonitoring has recently been explored as a practical method for examination of chemical exposure, however, there are few studies exploring the use of saliva for OP insecticides. To evaluate the use of salivary ChE as a biological monitor for OP insecticide exposure, a modified Ellman assay in conjunction with a pharmacodynamic model was used to characterize salivary ChE in adult male Sprague-Dawley rats. Comparison of rat saliva, brain, and plasma ChE activity in the presence of selective inhibitors of AChE and BuChE (BW284C51 and iso-OMPA, respectively) with different ChE substrates indicated that rat salivary ChE activity is primarily associated with BuChE (>95%). Further characterization of rat salivary BuChE kinetics yielded an average total BuChE active site concentration of 1.20+/-0.13 fmol ml(-1) saliva, an average reactivation rate constant (Kr) of 0.070+/-0.008 h(-1), and an inhibitory rate constant (Ki) of approximately 9 nM(-1) h(-1). The pharmacodynamic model successfully described the in vitro BuChE activity profile as well as the kinetic parameters. These results support the potential utility of saliva as a biomonitoring matrix for evaluating occupational and environmental exposure to CPF and other OP insecticides.