Merck
  • Eupatin, a Flavonoid, Inhibits Coronavirus 3CL Protease and Replication.
Eupatin, a Flavonoid, Inhibits Coronavirus 3CL Protease and Replication.
International journal of molecular sciences (2023-06-10)
Yea-In Park, Jang Hoon Kim, Siyun Lee, Ik Soo Lee, Junsoo Park
ABSTRACT
The coronavirus disease 2019 (COVID-19) pandemic has caused more than six million deaths worldwide since 2019. Although vaccines are available, novel variants of coronavirus are expected to appear continuously, and there is a need for a more effective remedy for coronavirus disease. In this report, we isolated eupatin from Inula japonica flowers and showed that it inhibits the coronavirus 3 chymotrypsin-like (3CL) protease as well as viral replication. We showed that eupatin treatment inhibits SARS-CoV-2 3CL-protease, and computational modeling demonstrated that it interacts with key residues of 3CL-protease. Further, the treatment decreased the number of plaques formed by human coronavirus OC43 (HCoV-OC43) infection and decreased viral protein and RNA levels in the media. These results indicate that eupatin inhibits coronavirus replication.