Merck
  • Emergence of immune escape at dominant SARS-CoV-2 killer T cell epitope.
Emergence of immune escape at dominant SARS-CoV-2 killer T cell epitope.
Cell (2022-08-06)
Garry Dolton, Cristina Rius, Md Samiul Hasan, Aaron Wall, Barbara Szomolay, Enas Behiry, Thomas Whalley, Joel Southgate, Anna Fuller, Théo Morin, Katie Topley, Li Rong Tan, Philip J R Goulder, Owen B Spiller, Pierre J Rizkallah, Lucy C Jones, Thomas R Connor, Andrew K Sewell
ABSTRACT
We studied the prevalent cytotoxic CD8 T cell response mounted against severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) Spike glycoprotein269-277 epitope (sequence YLQPRTFLL) via the most frequent human leukocyte antigen (HLA) class I worldwide, HLA A∗02. The Spike P272L mutation that has arisen in at least 112 different SARS-CoV-2 lineages to date, including in lineages classified as "variants of concern," was not recognized by the large CD8 T cell response seen across cohorts of HLA A∗02+ convalescent patients and individuals vaccinated against SARS-CoV-2, despite these responses comprising of over 175 different individual T cell receptors. Viral escape at prevalent T cell epitopes restricted by high frequency HLAs may be particularly problematic when vaccine immunity is focused on a single protein such as SARS-CoV-2 Spike, providing a strong argument for inclusion of multiple viral proteins in next generation vaccines and highlighting the need for monitoring T cell escape in new SARS-CoV-2 variants.
Sigma-Aldrich
Dasatinib, ≥98% (HPLC)
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Millipore
EDTA, Disodium Salt, Dihydrate, Molecular Biology Grade