Merck
  • Combined micro-osmotic pump infusion and intracerebroventricular injection to study FGF1 signaling pathways in the mouse brain.
Combined micro-osmotic pump infusion and intracerebroventricular injection to study FGF1 signaling pathways in the mouse brain.
STAR protocols (2022-04-29)
Jenny M Brown, Bao Anh Phan, Nadia Aalling, Gregory J Morton, Michael W Schwartz, Jarrad M Scarlett
ABSTRACT
Intracerebroventricular (icv) injection of fibroblast growth factor 1 (FGF1) elicits remission of diabetic hyperglycemia in rodent models of type 2 diabetes. Here, we present an optimized protocol to study the intracellular signaling pathways underlying the FGF1-induced sustained glucose lowering in the mouse brain. This protocol combines icv injection of FGF1 and osmotic mini-pump infusion of U0126, an inhibitor of MAPK/ERK signaling. We describe the surgical procedure and verification of U0126 inhibition of FGF1-stimulated hypothalamic MAPK/ERK signaling via western blot. For complete details on the use and execution of this protocol, please refer to Brown et al. (2021).
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.