Merck
  • Molecular mechanisms underlying the effects of the small molecule AMC-04 on apoptosis: Roles of the activating transcription factor 4-C/EBP homologous protein-death receptor 5 pathway.
Molecular mechanisms underlying the effects of the small molecule AMC-04 on apoptosis: Roles of the activating transcription factor 4-C/EBP homologous protein-death receptor 5 pathway.
Chemico-biological interactions (2020-10-03)
So Young Kim, Supyong Hwang, Min Kyung Choi, Sojung Park, Ky Youb Nam, Inki Kim
ABSTRACT
The unfolded protein response (UPR) is an emerging target pathway for cancer treatment owing to its ability to induce cell death. In our previous analysis of UPR-modulating small molecules, we had reported that piperazine oxalate derivative compounds (AMC-01-04) are able to promote increased phosphorylation of eukaryotic translation initiation factor-2 alpha (eIF2α). In this study, we found that AMC-04 induces apoptotic cell death via the activation of UPR in human breast and liver cancer cells. AMC-04 upregulated the expression of activating transcription factor-4 (ATF4)-C/EBP homologous protein (CHOP) and death receptor 5 (DR5) in cancer cells, as revealed by microarray analysis, small-interference RNA assay, and western blotting. From a mechanistic perspective, cytotoxic UPR pathway activation by AMC-04 is mediated by reactive oxygen species (ROS) and p38 mitogen-activated protein kinase (p38 MAPK) signaling. A chemical informatics approach predicted that AMC-04 modulates histone methyltransferase activity. Based on biochemical analysis, the activity of histone methyltransferases, including SUV39H1, SUV39H2, SETDB1, and EHMT1, was inhibited by AMC-04. Furthermore, chemical inhibition of the identified target proteins induced UPR activation and apoptotic cell death, suggesting that inhibition of histone methyltransferases is a promising strategy for cancer therapy. Taken together, we showed that the small molecule AMC-04 modulates epigenetic enzyme activity and mediates the link between cytotoxic UPR and histone modifications.
Sigma-Aldrich
UNC0642, ≥98% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human DDIT3