Merck
  • An RNA vaccine drives expansion and efficacy of claudin-CAR-T cells against solid tumors.
An RNA vaccine drives expansion and efficacy of claudin-CAR-T cells against solid tumors.
Science (New York, N.Y.) (2020-01-04)
Katharina Reinhard, Benjamin Rengstl, Petra Oehm, Kristina Michel, Arne Billmeier, Nina Hayduk, Oliver Klein, Kathrin Kuna, Yasmina Ouchan, Stefan Wöll, Elmar Christ, David Weber, Martin Suchan, Thomas Bukur, Matthias Birtel, Veronika Jahndel, Karolina Mroz, Kathleen Hobohm, Lena Kranz, Mustafa Diken, Klaus Kühlcke, Özlem Türeci, Ugur Sahin
ABSTRACT
Chimeric antigen receptor (CAR)-T cells have shown efficacy in patients with B cell malignancies. Yet, their application for solid tumors has challenges that include limited cancer-specific targets and nonpersistence of adoptively transferred CAR-T cells. Here, we introduce the developmentally regulated tight junction protein claudin 6 (CLDN6) as a CAR target in solid tumors and a strategy to overcome inefficient CAR-T cell stimulation in vivo. We demonstrate that a nanoparticulate RNA vaccine, designed for body-wide delivery of the CAR antigen into lymphoid compartments, stimulates adoptively transferred CAR-T cells. Presentation of the natively folded target on resident antigen-presenting cells promotes cognate and selective expansion of CAR-T cells. Improved engraftment of CAR-T cells and regression of large tumors in difficult-to-treat mouse models was achieved at subtherapeutic CAR-T cell doses.