Merck
  • Excitotoxicity is required for induction of oxidative stress and apoptosis in mouse striatum by the mitochondrial toxin, 3-nitropropionic acid.
Excitotoxicity is required for induction of oxidative stress and apoptosis in mouse striatum by the mitochondrial toxin, 3-nitropropionic acid.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2000-01-05)
G W Kim, J C Copin, M Kawase, S F Chen, S Sato, G T Gobbel, P H Chan
ABSTRACT
Excitotoxicity is implicated in the pathogenesis of several neurologic diseases, such as chronic neurodegenerative diseases and stroke. Recently, it was reported that excitotoxicity has a relationship to apoptotic neuronal death, and that the mitochondrial toxin, 3-nitropropionic acid (3-NP), could induce apoptosis in the striatum. Although striatal lesions produced by 3-NP could develop through an excitotoxic mechanism, the exact relationship between apoptosis induction and excitotoxicity after 3-NP treatment is still not clear. The authors investigated the role of excitotoxicity and oxidative stress on apoptosis induction within the striatum after intraperitoneal injection of 3-NP. The authors demonstrated that removal of the corticostriatal glutamate pathway reduced superoxide production and apoptosis induction in the denervated striatum of decorticated mice after 3-NP treatment. Also, the N-methyl-D-aspartate (NMDA) receptor antagonist, MK-801, prevented apoptosis in the striatum after 3-NP treatment for 5 days, whereas the non-NMDA receptor antagonist, 2,3-dihydroxy-6-nitro-7-sulphamoyl-benzo(F)quinoxaline, was ineffective. The authors also evaluated the initial type of neuronal death by 3-NP treatment for different durations from 1 to 5 days. In early striatal damage, apoptotic neuronal death initially occurred after 3-NP treatment. Our data show that excitotoxicity related to oxidative stress initially induces apoptotic neuronal death in mouse striatum after treatment with 3-NP.
Sigma-Aldrich
3-Nitropropionic acid, ≥97%