Merck
  • The inhibition of human leucocyte elastase and chymotrypsin-like protease by elastatinal and chymostatin.
The inhibition of human leucocyte elastase and chymotrypsin-like protease by elastatinal and chymostatin.
Biochimica et biophysica acta (1976-05-13)
G Feinstein, C J Malemud, A Janoff
ABSTRACT
The ability of elastatinal and chymostatin, protease inhibitors of microbial origin, to inhibit human leucocyte proteases (EC 3.4.-) was studied. Elastatinal and chymostatin are capable of inhibiting the pancreatic enzymes elastase and chymotrypsin, respectively. It was found in these studies, with synthetic substrates, that elastatinal is a much weaker inhibitor of human leucocyte elastase than it is of porcine pancreatic elastase. Elastatinal caused no inhibition of the activity of human leucocyte chymotrypsin-like protease. Chymostatin was found to be a powerful inhibitor of human leucocyte chymotrypsin-like protease. Its affinity to the leucocyte protease was higher than its affinity to bovine pancreatic alpha-chymotrypsin. Chymsotatin had a weak inhibitory effect on the activity of human leucocyte elastase. Studies were also carried out on the ability of chymostatin to inhibit the release of 35SO2-4 from rabbit articular cartilage by human leucocyte chymotrypsin-like protease. Preincubation of the chymostatin with the protease before the latter was added to the 35SO2-4 -labeled cartilage caused inhibition of proteolysis as measured by 35SO2-4 release. Preincubation of chymostatin with 35SO2-4 -labeled cartilage prior to addition of the human chymotrypsin-like protease to the tissue also inhibited 35SO2-4 release. However, in the case of preincubation of cartilage with alpha1 -antitrypsin there was no such inhibition. It therefore appeared that chymostatin, unlike alpha1 -antitrypsin, was capable of penetrating the cartilage matrix and exerting its inhibitory effect upon the human leucocyte chymotrypsin-like protease that was subsequently added to the tissue.