Skip to Content
Merck
Hh Signaling Antagonist X, Itraconazole The Hh Signaling Antagonist X, Itraconazole, also referenced under CAS 84625-61-6, controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Itraconazole

Hh Signaling Antagonist X, Itraconazole The Hh Signaling Antagonist X, Itraconazole, also referenced under CAS 84625-61-6, controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Cell Signaling applications.
Synonym(s):
Itraconazole, Oriconazole, R51211, Sporanox, Hh Signaling Antagonist X, Itraconazole
Empirical Formula (Hill Notation):
C35H38Cl2N8O4
CAS Number:
Molecular Weight:
705.63

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique