Tributyl 2-acetylcitrate
Synonym(s):
Acetyl tributyl citrate, Tributyl 2-acetylcitrate
Linear Formula:
CH3CO2C[CO2(CH2)3CH3][CH2CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
402.48
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3