Skip to Content
Merck
trans-4-Hydroxy-L-proline analytical standard

Hydroxyproline

trans-4-Hydroxy-L-proline analytical standard
Synonym(s):
trans-4-Hydroxy-L-proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, Hyp
Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
EC Number:

Products

Shipping
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2