Alumina
Synonym(s):
Aluminum oxide, Alumina
Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
Products
Shipping
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing