Skip to Content
Merck
vibrant-m

Ethylene glycol 5 M solution

vibrant-m
Synonym(s):
Ethylene glycol solution
Empirical Formula (Hill Notation):
C2H6O2
CAS Number:
Molecular Weight:
62.07

Products

Shipping
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2