Iodomethane solution
Synonym(s):
Methyl iodide
Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing