![Liothyronine for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/302/462/59c3b90b-f08f-4219-adc4-d18ddc021dfe/640/59c3b90b-f08f-4219-adc4-d18ddc021dfe.png)
Liothyronine for peak identification
![Liothyronine for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/302/462/59c3b90b-f08f-4219-adc4-d18ddc021dfe/640/59c3b90b-f08f-4219-adc4-d18ddc021dfe.png)
Synonym(s):
3,3′,5-Triiodo-L-thyronine sodium salt, Liothyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt, T3
Empirical Formula (Hill Notation):
C15H11I3NNaO4
CAS Number:
Molecular Weight:
672.96
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing