![Ethyl 4-(4,8-dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinecarboxylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/123/854/be2c3cb1-8963-4ec1-9e1f-4bbfec448cca/640/be2c3cb1-8963-4ec1-9e1f-4bbfec448cca.png)
Ethyl 4-(4,8-dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinecarboxylate
![Ethyl 4-(4,8-dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinecarboxylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/123/854/be2c3cb1-8963-4ec1-9e1f-4bbfec448cca/640/be2c3cb1-8963-4ec1-9e1f-4bbfec448cca.png)
Synonym(s):
4-Chloro-Loratadine
Empirical Formula (Hill Notation):
C22H22Cl2N2O2
CAS Number:
Molecular Weight:
417.33
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1