Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
AmberChrom
™
50WX4 hydrogen form
Linear Formula:
(C
10
H
12
· C
10
H
10
· C
8
H
8
)
x
CAS Number:
69011-20-7
Products
Shipping
Ships Today (2)
Product Category
resins (3)
Brand
Supelco (2)
Physical Form
Separation Technique
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
217484
AmberChrom
™
50WX4 hydrogen form,
200-400 mesh, Cation Exchange Resin
Expand
View Pricing
422096
AmberChrom
™
50WX4 hydrogen form,
hydrogen form, 100-200 mesh
Expand
View Pricing
428663
AmberChrom
™
50WX4 hydrogen form,
hydrogen form, 50-100 mesh
Expand
View Pricing