Skip to Content
Merck
Toluene suitable for HPLC, ≥99.8%

Toluene

Toluene suitable for HPLC, ≥99.8%
Synonym(s):
Toluene
Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique