4′-Hydroxy-3′-methoxyacetophenone
Synonym(s):
Acetovanillone, Apocynin
Linear Formula:
HOC6H3(OCH3)COCH3
CAS Number:
Molecular Weight:
166.17
Products
Shipping
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2