di-Sodium oxalate
Synonym(s):
di-Sodium oxalate
Empirical Formula (Hill Notation):
C2Na2O4
CAS Number:
Molecular Weight:
134.00
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing