Key Documents
502561
Upchurch Microtight® Tubing Sleeves
I.D. 0.007 in. (178 μm), yellow
About This Item
UNSPSC Code:
41115700
material
yellow
packaging
pkg of 10 ea
I.D.
0.007 in. (178 μm)
General description
Use these PEEK sleeves with all Microtight fittings, to connect capillary tubing. All are 0.025 in. O.D.; normally, choose an I.D. that is 0.002-0.003 in. larger than the O.D. of the capillary tubing. Color-coded.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Microtight is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)