Custom Peptides
- High quality peptide synthesis using FMOC and/or BOC chemistries
- Sequence length: 2-40mer
- Amount: up to 1 gram
- Purity Levels: Unpurified, 70%, 80%, 95%
- Comprehensive Modification offering including, FRET, C-terminal aldehyde, phosphorylation, multiple disulfide bonds, etc.
- Format: Supplied lyophilized
- QC Analysis: All peptides by HPLC and MS; additional analysis available upon request
- Learn more