- Inhibition of SNK-SPAR signaling pathway promotes the restoration of motor function in a rat model of ischemic stroke.
Inhibition of SNK-SPAR signaling pathway promotes the restoration of motor function in a rat model of ischemic stroke.
Journal of cellular biochemistry (2017-07-12)
Qing-Yu Fan, Jing-Jie Liu, Gui-Lian Zhang, Hai-Qin Wu, Ru Zhang, Shu-Qin Zhan, Na Liu
PMID28696012
ABSTRACT
This study aimed to investigate the effects of SPAR signaling pathway on the restoration of motor function in ischemic stroke (IS). Sprague-Dawley male rats were separated into the control and sham groups, as well as the group for middle cerebral artery occlusion (MCAO) model establishment. Successfully established rat ischemic models were randomly divided into model, SNK