Merck
  • Occurrence and fate of substituted diphenylamine antioxidants and benzotriazole UV stabilizers in various Canadian wastewater treatment processes.
Occurrence and fate of substituted diphenylamine antioxidants and benzotriazole UV stabilizers in various Canadian wastewater treatment processes.
Water research (2017-08-02)
Zhe Lu, Shirley Anne Smyth, Thomas E Peart, Amila O De Silva
ABSTRACT
Substituted diphenylamine antioxidants (SDPAs) and benzotriazole UV stabilizers (BZT-UVs) are additives used in industrial and consumer products to prevent degradation or color change of materials, but their environmental fate and disposition are not well characterized. In this study, SDPAs and BZT-UVs were analyzed in 68 liquid and 39 solid samples collected from 9 wastewater treatment plants (WWTPs) in Canada to investigate the occurrence and fate of these contaminants. The median concentrations of ΣSDPAs and ΣBZT-UVs was 483 and 76.2 ng L
Sigma-Aldrich
2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-4-methylphenol, 98%